Kemisk øse
Kemisk øse, PP, 1000 ml, 1000 mm
331-2112
: BURKLE
Liquid samplers for aggressive liquids, with fixed rod.
- Three sizes
- Entirely made of polypropylene − No metal parts
- Parts coming into contact with sample media made of PP
- Kapacitet:1000 ml
- Beskrivelse:Kemisk øse
- Længde:1000 mm
- Materiale:PP