Kemisk øse
Kemisk øse, PP, 500 ml, 1000 mm
Catalog # 331-2111
Supplier: BURKLE
Gå til:
Product Details & Documents
Liquid samplers for aggressive liquids, with fixed rod.
- Three sizes
- Entirely made of polypropylene − No metal parts
- Parts coming into contact with sample media made of PP
Specifikationer
- Kapacitet:500 ml
- Beskrivelse:Kemisk øse
- Længde:1000 mm
- Materiale:PP
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



