Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Double bossheads

Double bossheads

Double bosshead, safety heart screw M8, aluminium, 16,5 mm
Katalog # 241-3177
Leverandør:   BOCHEM
Double bossheads
Katalog # 241-3177
Leverandør:  BOCHEM
Double bossheads
Double bossheads

Double bosshead, safety heart screw M8, aluminium, 16,5 mm

Price
181,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Double bossheads, either made of aluminium or malleable cast iron, chromium plated.

  • For rods up to 16,5 mm
  • Angle: 90°
  • With safety screw type 'heart' (M8)

 : DIN 12895

Specifikationer
  • Beskrivelse:
    Double bosshead, safety heart screw M8
  • Materiale:
    aluminium
  • Seq:
    01
  • Til stang Ø:
    16,5 mm