Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
185 mm, 6 mm
Katalog # 231-2323
Leverandør:   RSG
Double spatulas, micro
Katalog # 231-2323
Leverandør:  RSG
Double spatulas, micro
Double spatulas, micro
185 mm, 6 mm
Price
311,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Stainless steel. Anti-magnetic, very flexible micro powder spatula.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Documents
Specifikationer
  • Længde:
    185 mm
  • Bladbredde:
    6 mm