Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Knife spatulas, angled

Knife spatulas, angled

230 mm, 130 mm
Katalog # 231-2320
Leverandør:   HAMMACHER KARL
Knife spatulas, angled
Katalog # 231-2320
Leverandør:  HAMMACHER KARL
Knife spatulas, angled
Knife spatulas, angled

230 mm, 130 mm

Price
495,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Flexible knife spatulas with wooden handle and angled blade made of stainless steel.

Specifikationer
  • Længde:
    230 mm
  • Seq:
    13
  • Klinge længde:
    130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.