Powder spatulas, micro
140 mm, 4 mm
Katalog # 231-2316
Leverandør: HAMMACHER KARL
Gå til:
Product Details & Documents
Single-ended powder spatulas made from WIRONIT steel.
Specifikationer
- Længde:140 mm
- Bladbredde:4 mm