Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Spatulas, micro

Spatulas, micro

140 mm, 3 mm
Catalog # HAMMHWN002-03
Supplier:   HAMMACHER KARL
Spatulas, micro
Catalog # HAMMHWN002-03
Supplier:  HAMMACHER KARL
Spatulas, micro
Spatulas, micro

140 mm, 3 mm

Price
382,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Single-ended micro spatulas made from WIRONIT steel.

Specifications
  • Længde:
    140 mm
  • Seq:
    02
  • Bladbredde:
    3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.