Orders LinkContactUsLinkComponent
Filter discs
Filter discs
76,2 mm, 3,18 mm
Katalog # 42301.KS
Leverandør:   Thermo Fisher Scientific
Filter discs
Katalog # 42301.KS
Filter discs
76,2 mm, 3,18 mm
Price
1.650,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These quartz discs are manufactured by fusing naturally occurring crystalline silica.
  • Ground and polished
Specifikationer
  • Ø:
    76,2 mm
  • Seq:
    04
  • Tykkelse:
    3,18 mm