n-Hexan ≥95%, Environmental Grade for environmental analysis
4x1 L, 4
Leverandør: Thermo Fisher Scientific
n-Hexan ≥95%, Environmental Grade for environmental analysis
Katalog # 42100.M1
Leverandør: Thermo Fisher Scientific
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Purity: 95+% CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520
Pesticide residue, environmental and GC analysis
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:14,4694
- Kogende Pt:69 °C
- Smeltende Pt:–95 °C
- Tæthed:0,659
- Flash Pt:–23 °C (–9F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Documents
Specifikationer
Specifikationer
- Pakn:4
- Pk Info:4x1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



