n-Hexan ≥95%, Environmental Grade for environmental analysis
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
n-Hexan ≥95%, Environmental Grade for environmental analysis
Katalog # 42100.K2
Leverandør: Thermo Fisher Scientific
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Purity: 95+% CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520
Pesticide residue, environmental and GC analysis
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:14,4694
- Kogende Pt:69 °C
- Smeltende Pt:–95 °C
- Tæthed:0,659
- Flash Pt:–23 °C (–9F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L



