Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Threaded fittings

Nipple, with outer thread, NW 23,5 mm, PP, 25,4 mm
Katalog # 223-1151
Leverandør:   BURKLE
Threaded fittings
Katalog # 223-1151
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Nipple, with outer thread, NW 23,5 mm, PP, 25,4 mm

Price
56,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    25,4 mm
  • Beskrivelse:
    Nipple, with outer thread, NW 23,5 mm
  • Materiale:
    PP
  • Seq:
    41