Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

T-piece, with inner thread, NW 23 mm, PP, 19,05 mm
Catalog # 223-1148
Supplier:   BURKLE
Threaded fittings
Catalog # 223-1148
Supplier:  BURKLE
Threaded fittings
Threaded fittings

T-piece, with inner thread, NW 23 mm, PP, 19,05 mm

Price
72,20 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Gevind:
    19,05 mm
  • Beskrivelse:
    T-piece, with inner thread, NW 23 mm
  • Materiale:
    PP
  • Seq:
    32
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.