Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Threaded fittings

Tubing nozzle, fix, bend, external thread, nozzle-Ø 25 mm, NW 19,5 mm, PP, 25,4 mm
  223-1145
 :   BURKLE
Threaded fittings
  223-1145
 :  BURKLE
Threaded fittings
Threaded fittings

Tubing nozzle, fix, bend, external thread, nozzle-Ø 25 mm, NW 19,5 mm, PP, 25,4 mm

 
44,30 kr
   
1 * 1 Items
 
 
  • Gevind:
    25,4 mm
  • Beskrivelse:
    Tubing nozzle, fix, bend, external thread, nozzle-Ø 25 mm, NW 19,5 mm
  • Materiale:
    PP
  • Seq:
    24
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.