Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Threaded fittings

Tubing nozzles, fixed, external thread, nozzle-Ø 16 mm, NW 10,5 mm, PP, 19,05 mm
Katalog # 223-1144
Leverandør:   BURKLE
Threaded fittings
Katalog # 223-1144
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Tubing nozzles, fixed, external thread, nozzle-Ø 16 mm, NW 10,5 mm, PP, 19,05 mm

Price
20,20 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    19,05 mm
  • Beskrivelse:
    Tubing nozzles, fixed, external thread, nozzle-Ø 16 mm, NW 10,5 mm
  • Materiale:
    PP