Orders LinkContactUsLinkComponent
Stoppers, LDPE
Stoppers, LDPE
ufarvet, 11,5 mm, 13 mm
Katalog # 217-9249
Leverandør:   SEMADENI
Stoppers, LDPE
Katalog # 217-9249
Leverandør:  SEMADENI
Stoppers, LDPE
Stoppers, LDPE
ufarvet, 11,5 mm, 13 mm
Price
kr98.10
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Lamellar stoppers made from soft PE-LD.
  • Practical grip edge
  • Bright tolerance through soft lamellars
Documents
Specifikationer
  • Ø bund:
    11,5 mm
  • Ø top:
    13 mm
  • Farve:
    ufarvet
  • Seq:
    05