Orders LinkContactUsLinkComponent
Caps for tubes
Caps for tubes
Cap for 12 and 13 mm tubes, ufarvet
Catalog # 217-8932
Supplier:   DELTALAB
Caps for tubes
Catalog # 217-8932
Supplier:  DELTALAB
Caps for tubes
Cap for 12 and 13 mm tubes, ufarvet
Price
441,00 kr
Log in for availability
1 * 2.000 Items
Product Details & Documents
Polyethylene caps for all types of polypropylene and polystyrene tubes.
Specifications
  • Farve:
    ufarvet
  • Beskrivelse:
    Cap for 12 and 13 mm tubes
  • Seq:
    10