Wire baskets, rectangular, with handle
With dividers, 4 compartments, 480×300×150 mm
Katalog # 216-7515
Leverandør: DELTALAB
Gå til:
Product Details & Documents
Made of stainless steel.
- Features a strong woven mesh base
- With folding handles
- Suitable for bottles and containers
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Beskrivelse:With dividers, 4 compartments
- Seq:02
- B×D×H:480×300×150 mm