Wrench for jerricans
Wrench for jerricans, HDPE, 45 mm
Katalog # 216-5571
Leverandør: Kautex
Gå til:
Product Details & Documents
Wrenches made from HDPE for opening and closing jerricans.
Specifikationer
- Kæbestørrelse:45 mm
- Beskrivelse:Wrench for jerricans
- Materiale:HDPE
- Seq:01