Jars with overlapping lid
10 ml, 40 mm, 20 mm
Katalog # 216-3155
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Jars, made of PP, with overlapping lid.
- Lids can be labelled
- Suitable for ointments, powders, and granules
- Good chemical and temperature resistance
Specifikationer
- Udv.Ø:40 mm
- Kapacitet:10 ml
- Højde:20 mm