Instrument dishes
Låg til 216-1405 skåle, 275x150x10 mm
Catalog # 216-1415
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Thick-walled glass.
- Can be sterilised in hot air up to 180 °C
- Robust
- Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifications
- Beskrivelse:Låg til 216-1405 skåle
- Seq:12
- B×D×H:275x150x10 mm