Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Bowls, PP

13000 ml, 180 mm, 400 mm
  216-1207
 :   VITLAB
Bowls, PP
  216-1207
 :  VITLAB
Bowls, PP
Bowls, PP

13000 ml, 180 mm, 400 mm

 
178,00 kr
   
1 * 1 Items
 

Bowls, made of white PP.

  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean

Ideal for safe storage of food products. With broad, stable edge and circumferential standing ring on the bottom.

 
  • Udv.Ø:
    400 mm
  • Kapacitet:
    13000 ml
  • Højde:
    180 mm
  • Seq:
    07