Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-1-Methoxy-2-propanol ≥99%

(±)-1-Methoxy-2-propanol ≥99%

Glass bottle, 4x4 L, 4
Katalog # 41547.M6
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
(±)-1-Methoxy-2-propanol ≥99%
Katalog # 41547.M6
CAS nummer:  107-98-2
(±)-1-Methoxy-2-propanol ≥99%
(±)-1-Methoxy-2-propanol ≥99%

Glass bottle, 4x4 L, 4

CAS nummer:  107-98-2
Price
5.080,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents

Purity: 99+% CAS No.: 107-98-2 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 0.926 Refractive Index: 1.4030 Appearance: Colourless Odour: Alcohol-like pH: 4-7 Melting Point: -100° Boiling Point: 119-120° Beilstein Reference: 1731270 UN No.: 3092 MDL No.: MFCD00004537

Synonymer:  (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol
  • CAS nummer:107-98-2
  • Formel:CH₃CH(OH)CH₂OCH₃
  • MDL nummer:MFCD00004537
  • Molecular Weight:90.12 g/mol
  • Kogende Pt:119…120 °C
  • Smeltende Pt:–100 °C
  • Tæthed:0,926
  • Flash Pt:35 °C (95 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-539-1
  • FN:3092
  • ADR:3,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    4
  • Pk Info:
    4x4 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.