Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(±)-1-Methoxy-2-propanol ≥99%

Glass bottle, -, 4 L
Katalog # 41547.K7
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
(±)-1-Methoxy-2-propanol ≥99%
Katalog # 41547.K7
CAS nummer:  107-98-2
(±)-1-Methoxy-2-propanol ≥99%
(±)-1-Methoxy-2-propanol ≥99%

Glass bottle, -, 4 L

CAS nummer:  107-98-2
Price
1.540,00 kr
Log ind for tilgængelighed
1 * 4 L
Product Details & Documents

Purity: 99+% CAS No.: 107-98-2 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 0.926 Refractive Index: 1.4030 Appearance: Colourless Odour: Alcohol-like pH: 4-7 Melting Point: -100° Boiling Point: 119-120° Beilstein Reference: 1731270 UN No.: 3092 MDL No.: MFCD00004537

Synonymer:  (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol
  • CAS nummer:107-98-2
  • Formel:CH₃CH(OH)CH₂OCH₃
  • MDL nummer:MFCD00004537
  • Molecular Weight:90.12 g/mol
  • Kogende Pt:119…120 °C
  • Smeltende Pt:–100 °C
  • Tæthed:0,926
  • Flash Pt:35 °C (95 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-539-1
  • FN:3092
  • ADR:3,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    4 L
  • Pk Info:
    -

Specifikationer