Orders LinkContactUsLinkComponent
Laboratory bottles, narrow neck, with thread
Laboratory bottles, narrow neck, with thread
20000 ml, Bottle with plastic coating, GL 45
Katalog # 215-1562
Leverandør:   witeg Labortechnik
Laboratory bottles, narrow neck, with thread
Katalog # 215-1562
Leverandør:  witeg Labortechnik
Laboratory bottles, narrow neck, with thread
Laboratory bottles, narrow neck, with thread
20000 ml, Bottle with plastic coating, GL 45
Price
8.250,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These laboratory bottles are made of borosilicate glass with safety coating witofoam.
  • Excellent chemical resistance, good heat resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Compressive strength is not increased by the plastic coating
Round, with graduation and DIN thread. The coating provides scratch, leak and splinter protection and is ideally suited to both the transport and storage of hazardous media or valuable samples.
Specifikationer
  • Gevind:
    GL 45
  • Kapacitet:
    20000 ml
  • Beskrivelse:
    Bottle with plastic coating