Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Jars, with snap-on lid

Jars, with snap-on lid

100 ml, 47 mm, 75 mm
Katalog # 215-0298
Leverandør:   SEMADENI
Jars, with snap-on lid
Katalog # 215-0298
Leverandør:  SEMADENI
Jars, with snap-on lid
Jars, with snap-on lid

100 ml, 47 mm, 75 mm

Price
2.810,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Jars, made of PET, with PE snap-on lid.

Ideal for the storage of granules, ointments, pastes and powders.

Specifikationer
  • Udv.Ø:
    47 mm
  • Kapacitet:
    100 ml
  • Højde:
    75 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.