Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Jars, with snap-on lid

50 ml, 40 mm, 60 mm
Katalog # 215-0297
Leverandør:   SEMADENI
Jars, with snap-on lid
Katalog # 215-0297
Leverandør:  SEMADENI
Jars, with snap-on lid
Jars, with snap-on lid

50 ml, 40 mm, 60 mm

Price
2.530,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Jars, made of PET, with PE snap-on lid.

Ideal for the storage of granules, ointments, pastes and powders.

Specifikationer
  • Udv.Ø:
    40 mm
  • Kapacitet:
    50 ml
  • Højde:
    60 mm