Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
1000 ml, 22 mm, 75×85×210 mm
Katalog # 215-0240
Leverandør: SEMADENI
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Katalog # 215-0240
Leverandør: SEMADENI
Gå til:
Product Details & Documents
HDPE, translucent, with thread.
- Good chemical resistance
- Good temperature resistance
Ordering information: Screw caps must be ordered separately.
Specifikationer
- Kapacitet:1000 ml
- B×D×H:75×85×210 mm
- Hals indv. Ø:22 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



