Push-in caps for tubes
LDPE, Caps, Ø 46 mm, ufarvet
Katalog # 215-0217
Gå til:
Discontinued
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifikationer
- Farve:ufarvet
- Beskrivelse:Caps, Ø 46 mm
- Materiale:LDPE
- Seq:22