Measuring beakers with handle
1000 ml, 167 mm, 125 mm, 10 ml
Katalog # 213-3746
Leverandør: VITLAB
Gå til:
Product Details & Documents
PP, meget gennemskinnelig.
- Volume is equally visible for left and right handers
- Good chemical resistance, resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
- To preserve markings, cleaning at no higher than +60 °C is recommended
- For autoclaving we recommend the design with raised graduations
Ideal for general laboratory use and a wide variety of other applications. With stable handle, spout and easily readable, printed black scale on both sides. Stackable.
: Food-safe products acc. to EC Directive No. 10/2011.
Specifikationer
- Udv.Ø:125 mm
- Inddelings-interval:10 ml
- Kapacitet:1000 ml
- Højde:167 mm
- Seq:03