Test tube racks
17 mm, 50 (10×5), 220×110×50 mm
Catalog # 212-8803
Supplier: DELTALAB
Jump to:
Product Details & Documents
Test tube racks, made of aluminium or stainless steel.
- Stackable
- Alpha-numerical index for better sample location
- Good chemical and temperature resistance
Delivery information: Supplied encased and interlocked, wrapped in a protecting film.
Documents
Certificates
Search by Lot NumberSpecifications
- Hul Ø:17 mm
- Ant. huller:50 (10×5)
- Seq:12
- B×D×H:220×110×50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



