Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Test tube racks

12 mm, 50 (10×5), 165×85×50 mm
Katalog # 212-8801
Leverandør:   DELTALAB
Test tube racks
Katalog # 212-8801
Leverandør:  DELTALAB
Test tube racks

12 mm, 50 (10×5), 165×85×50 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Test tube racks, made of aluminium or stainless steel.

  • Stackable
  • Alpha-numerical index for better sample location
  • Good chemical and temperature resistance

 : Supplied encased and interlocked, wrapped in a protecting film.

 

Specifikationer
  • Hul Ø:
    12 mm
  • Ant. huller:
    50 (10×5)
  • Seq:
    10
  • B×D×H:
    165×85×50 mm