Test tube racks
12 mm, 50 (10×5), 165×85×45 mm
Katalog # 212-8401
Leverandør: DELTALAB
Gå til:
Product Details & Documents
Test tube racks, made of aluminium or stainless steel.
- Stackable
- Alpha-numerical index for better sample location
- Good chemical and temperature resistance
Leveringsinformation: Supplied encased and interlocked, wrapped in a protecting film.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Hul Ø:12 mm
- Ant. huller:50 (10×5)
- Seq:04
- B×D×H:165×85×45 mm
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



