Reagent glassware cleaner
Test tube wiper, spade shape, rubber, 36×33 mm, rød
212-7218
Natural rubber or silicone.
- With hole for rods with 5/6 mm diameter
- Soft and flexible
- In 3 models available
- Farve:rød
- Beskrivelse:Test tube wiper, spade shape, rubber, 36×33 mm
- Seq:04