Reagent glassware cleaner
Test tube wiper, spade shape, rubber, 36×33 mm, grå
Katalog # 212-7216
Leverandør: DEUTSCH NEUMANN
Gå til:
Product Details & Documents
Natural rubber or silicone.
- With hole for rods with 5/6 mm diameter
- Soft and flexible
- In 3 models available
Specifikationer
- Farve:grå
- Beskrivelse:Test tube wiper, spade shape, rubber, 36×33 mm
- Seq:03