Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for Assortment Boxes, PS

Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, green, 162×108 mm, 45 mm height
Katalog # 211-2743
Leverandør:   SEMADENI
Accessories for Assortment Boxes, PS
Katalog # 211-2743
Leverandør:  SEMADENI
Accessories for Assortment Boxes, PS
Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, green, 162×108 mm, 45 mm height

Price
70,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Compartments, PS for storage boxes, green, 162×108 mm, 45 mm height
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.