Orders LinkContactUsLinkComponent
Screwthread tubes, attachable
Screwthread tubes, attachable
GL 14, 12 mm, 100 mm
Katalog # 201-1601
Leverandør:   witeg Labortechnik
Screwthread tubes, attachable
Katalog # 201-1601
Leverandør:  witeg Labortechnik
Screwthread tubes, attachable
Screwthread tubes, attachable
GL 14, 12 mm, 100 mm
Price
21,40 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
Specifikationer
  • Gevind:
    GL 14
  • Rør udv. Ø:
    12 mm
  • Seq:
    01
  • Samlet længde:
    100 mm
  • Cat. No.:
    201-1601