Orders LinkContactUsLinkComponent
Natriumiodid ≥99.5% ACS
Glass bottle for solids, 100 g
  40196.22
Natriumiodid ≥99.5% ACS
  40196.22
 :  7681-82-5
Natriumiodid ≥99.5% ACS
Natriumiodid ≥99.5% ACS
Glass bottle for solids, 100 g
 :  7681-82-5
853,00 kr
1 * 100 g
Purity: 99.5% min CAS No.: 7681-82-5 Molecular Formula: NaI Formula Weight: 149.89 Physical Form: Crystalline Powder Density: 3.67 Refractive Index: 1.7745 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 651° Boiling Point: 1300-1304° Solubility: Soluble in water, alcohol, glycerol, acetone. Insoluble in ether Merck Reference: 14,8631 MDL No.: MFCD00003532
 :  Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
  •  :7681-82-5
  •  :NaI
  •  :MFCD00003532
  •  :149.89 g/mol
  •  :14,8631
  •  :1300…1304 °C
  •  :651 °C
  •  :3,67
  •  :Stue temperatur
  •  :231-679-3
  •  :3077
  •  :9,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g