Natriumiodid ≥99.5% ACS
Glass bottle for solids, 100 g
Purity: 99.5% min CAS No.: 7681-82-5 Molecular Formula: NaI Formula Weight: 149.89 Physical Form: Crystalline Powder Density: 3.67 Refractive Index: 1.7745 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 651° Boiling Point: 1300-1304° Solubility: Soluble in water, alcohol, glycerol, acetone. Insoluble in ether Merck Reference: 14,8631 MDL No.: MFCD00003532
: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
- :7681-82-5
- :NaI
- :MFCD00003532
- :149.89 g/mol
- :14,8631
- :1300…1304 °C
- :651 °C
- :3,67
- :Stue temperatur
- :231-679-3
- :3077
- :9,III
- Pakning:Glass bottle for solids
- Pakn:100 g