Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumdihydrogenphosphat ≥96%

Glass bottle for solids, 1 kg
Katalog # 40145.A1
Irriterende
Leverandør:   Thermo Fisher Scientific
Natriumdihydrogenphosphat ≥96%
Katalog # 40145.A1
CAS nummer:  7558-80-7
Natriumdihydrogenphosphat ≥96%
Natriumdihydrogenphosphat ≥96%

Glass bottle for solids, 1 kg

CAS nummer:  7558-80-7
Price
807,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

Purity: 96% CAS No.: 7558-80-7 Molecular Formula: H2NaO4P Formula Weight: 119.98 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,8660 MDL No.: MFCD00003527

Synonymer:  Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
  • CAS nummer:7558-80-7
  • Formel:NaH₂PO₄
  • MDL nummer:MFCD00003527
  • Molecular Weight:119.98 g/mol
  • Merck Index:14,8660
  • Smeltende Pt:∼650 °C
  • Tæthed:0,9996 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-449-2
  • NÅ:01-2119489796-13
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.