Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Calciumstearat

Glass bottle for solids, 5 kg
Katalog # 39423.A7
Leverandør:   Thermo Fisher Scientific
Calciumstearat
Katalog # 39423.A7
CAS nummer:  1592-23-0
Calciumstearat
Calciumstearat

Glass bottle for solids, 5 kg

CAS nummer:  1592-23-0
Price
2.430,00 kr
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents

CAS No.: 1592-23-0 Molecular Formula: Ca[CH3(CH2)16CO2]2 Formula Weight: 607.04 Physical Form: Powder Odour: Odourless Melting Point: 179-180° MDL No.: MFCD00036390

Waterproofing and release agent, stabilizer for PVC, lubricant, conditioning agent

Synonymer:  Stearic acid calcium salt
  • CAS nummer:1592-23-0
  • Formel:C₃₆H₇₀CaO₄
  • MDL nummer:MFCD00036390
  • Molecular Weight:607.03 g/mol
  • Merck Index:14,1703
  • Smeltende Pt:179…180 °C
  • Tæthed:1,065 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:216-472-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 kg

Specifikationer