Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Calciumstearat

Calciumstearat

Glass bottle for solids, 1 kg
Katalog # 39423.A1
Leverandør:   Thermo Fisher Scientific
Calciumstearat
Katalog # 39423.A1
CAS nummer:  1592-23-0
Calciumstearat
Calciumstearat

Glass bottle for solids, 1 kg

CAS nummer:  1592-23-0
Price
707,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

CAS No.: 1592-23-0 Molecular Formula: Ca[CH3(CH2)16CO2]2 Formula Weight: 607.04 Physical Form: Powder Odour: Odourless Melting Point: 179-180° MDL No.: MFCD00036390

Waterproofing and release agent, stabilizer for PVC, lubricant, conditioning agent

Synonymer:  Stearic acid calcium salt
  • CAS nummer:1592-23-0
  • Formel:C₃₆H₇₀CaO₄
  • MDL nummer:MFCD00036390
  • Molecular Weight:607.03 g/mol
  • Merck Index:14,1703
  • Smeltende Pt:179…180 °C
  • Tæthed:1,065 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:216-472-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.