Methylethylketon 99+% ACS
Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Methylethylketon 99+% ACS
Katalog # 39119.AP
Leverandør: Thermo Fisher Scientific
CAS nummer: 78-93-3
Gå til:
Product Details & Documents
Purity: 99+% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.805 Refractive Index: 1.3790 Odour: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648
Synonymer: Ethylmethylketon, Butanon
- CAS nummer:78-93-3
- Formel:C₂H₅COCH₃
- MDL nummer:MFCD00011648
- Molecular Weight:72.11 g/mol
- Merck Index:14,6072
- Kogende Pt:80 °C
- Smeltende Pt:–87 °C
- Tæthed:0,805
- Flash Pt:–3 °C (26 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-159-0
- FN:1193
- ADR:3,II
- NÅ:01-2119457290-43
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
- Cat. No.:39119.AP
- Miljøcertificering:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



