Methylrød ACS
Glass bottle for solids, 100 g
CAS No.: 493-52-7 Molecular Formula: C15H15N3O2 Formula Weight: 269.30 Physical Form: Crystalline Appearance: Dark red Melting Point: 179-182° Beilstein Reference: 750102 Merck Reference: 14,6119 MDL No.: MFCD00002425
pH indicator in pH range 4.2 - 6.3
: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- :493-52-7
- :C₁₅H₁₅N₃O₂
- :MFCD00002425
- :269.3 g/mol
- :14,6119
- :179…182 °C
- :1,31 g/cm³ (20 °C)
- :Stue temperatur
- :207-776-1
Documents
- Pakning:Glass bottle for solids
- Pakn:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



