Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylrød ACS

Methylrød ACS

Glass bottle for solids, 25 g
Katalog # 36682.14
Leverandør:   Thermo Fisher Scientific
Methylrød ACS
Katalog # 36682.14
CAS nummer:  493-52-7
Methylrød ACS
Methylrød ACS

Glass bottle for solids, 25 g

CAS nummer:  493-52-7
Price
489,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

CAS No.: 493-52-7 Molecular Formula: C15H15N3O2 Formula Weight: 269.30 Physical Form: Crystalline Appearance: Dark red Melting Point: 179-182° Beilstein Reference: 750102 Merck Reference: 14,6119 MDL No.: MFCD00002425

pH indicator in pH range 4.2 - 6.3

Synonymer:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS nummer:493-52-7
  • Formel:C₁₅H₁₅N₃O₂
  • MDL nummer:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:14,6119
  • Smeltende Pt:179…182 °C
  • Tæthed:1,31 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:207-776-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer