Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Glycin ≥99%

Glycin ≥99%

Glass bottle for solids, 1 kg
Katalog # 36435.A1
Leverandør:   Thermo Fisher Scientific
Glycin ≥99%
Katalog # 36435.A1
CAS nummer:  56-40-6
Glycin ≥99%
Glycin ≥99%

Glass bottle for solids, 1 kg

CAS nummer:  56-40-6
Price
1.560,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

Purity: 99.5+% CAS No.: 56-40-6 Molecular Formula: H2NCH2CO2H Formula Weight: 75.07 Physical Form: Powder Density: 1.595 pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131

Synonymer:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS nummer:56-40-6
  • Formel:NH₂CH₂COOH
  • MDL nummer:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:14,4491
  • Kogende Pt:181 °C (1013 hPa)
  • Smeltende Pt:∼ 245 °C (dec.)
  • Tæthed:1,595
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-272-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.