Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Glycin ≥99%

Glass bottle for solids, 250 g
Katalog # 36435.30
Leverandør:   Thermo Fisher Scientific
Glycin ≥99%
Katalog # 36435.30
CAS nummer:  56-40-6
Glycin ≥99%
Glycin ≥99%

Glass bottle for solids, 250 g

CAS nummer:  56-40-6
Price
565,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 99.5+% CAS No.: 56-40-6 Molecular Formula: H2NCH2CO2H Formula Weight: 75.07 Physical Form: Powder Density: 1.595 pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131

Synonymer:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS nummer:56-40-6
  • Formel:NH₂CH₂COOH
  • MDL nummer:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:14,4491
  • Kogende Pt:181 °C (1013 hPa)
  • Smeltende Pt:∼ 245 °C (dec.)
  • Tæthed:1,595
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-272-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer