Hydroxylammoniumchlorid ≥96% ACS
Glass bottle for solids, 500 g
Purity: 96+% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Physical Form: Solid Density: 1.67 Appearance: White Odour: Acidic pH: 3.6 Melting Point: 152° dec. Solubility: Soluble in water, alcohol, methanol, glycerol Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089
A monoamine oxidase inhibitor; inhibits platelet aggregation
: Hydroxylamine HCl, Hydroxylamine hydrochloride
- :5470-11-1
- :NH₂OH·HCl
- :MFCD00051089
- :69.49 g/mol
- :14,4828
- :306 °C (1013 hPa)
- :152 °C (dec.)
- :1,67
- :Stue temperatur
- :226-798-2
- :2923
- :8,III
- Pakning:Glass bottle for solids
- Pakn:500 g