Orders LinkContactUsLinkComponent
Hydroxylammoniumchlorid ≥96% ACS
Glass bottle for solids, 500 g
  36416.36
Hydroxylammoniumchlorid ≥96% ACS
  36416.36
 :  5470-11-1
Hydroxylammoniumchlorid ≥96% ACS
Hydroxylammoniumchlorid ≥96% ACS
Glass bottle for solids, 500 g
 :  5470-11-1
2.540,00 kr
1 * 500 g
Purity: 96+% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Physical Form: Solid Density: 1.67 Appearance: White Odour: Acidic pH: 3.6 Melting Point: 152° dec. Solubility: Soluble in water, alcohol, methanol, glycerol Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089
A monoamine oxidase inhibitor; inhibits platelet aggregation
 :  Hydroxylamine HCl, Hydroxylamine hydrochloride
  •  :5470-11-1
  •  :NH₂OH·HCl
  •  :MFCD00051089
  •  :69.49 g/mol
  •  :14,4828
  •  :306 °C (1013 hPa)
  •  :152 °C (dec.)
  •  :1,67
  •  :Stue temperatur
  •  :226-798-2
  •  :2923
  •  :8,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g