Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Hydroxylammoniumchlorid ≥96% ACS

Hydroxylammoniumchlorid ≥96% ACS

Plastic bottle for solids, 100 g
Katalog # 36416.22
Sundhed
Ætsende
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
Hydroxylammoniumchlorid ≥96% ACS
Katalog # 36416.22
CAS nummer:  5470-11-1
Hydroxylammoniumchlorid ≥96% ACS
Hydroxylammoniumchlorid ≥96% ACS

Plastic bottle for solids, 100 g

CAS nummer:  5470-11-1
Price
630,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 96+% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Physical Form: Solid Density: 1.67 Appearance: White Odour: Acidic pH: 3.6 Melting Point: 152° dec. Solubility: Soluble in water, alcohol, methanol, glycerol Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089

A monoamine oxidase inhibitor; inhibits platelet aggregation

Synonymer:  Hydroxylamine HCl, Hydroxylamine hydrochloride
  • CAS nummer:5470-11-1
  • Formel:NH₂OH·HCl
  • MDL nummer:MFCD00051089
  • Molecular Weight:69.49 g/mol
  • Merck Index:14,4828
  • Kogende Pt:306 °C (1013 hPa)
  • Smeltende Pt:152 °C (dec.)
  • Tæthed:1,67
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:226-798-2
  • FN:2923
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.