Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumthiosulfat standardised solution 0.01 N in aqueous solution

Natriumthiosulfat standardised solution 0.01 N in aqueous solution

Plastic bottle, 1 L
Katalog # 35643.K2
Leverandør:   Thermo Fisher Scientific
Natriumthiosulfat standardised solution 0.01 N in aqueous solution
Katalog # 35643.K2
CAS nummer:  7772-98-7
Natriumthiosulfat standardised solution 0.01 N in aqueous solution
Natriumthiosulfat standardised solution 0.01 N in aqueous solution

Plastic bottle, 1 L

CAS nummer:  7772-98-7
Price
447,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Physical Form: Liquid MDL No.: MFCD00003499

Synonymer:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS nummer:7772-98-7
  • Formel:Na₂S₂O₃
  • MDL nummer:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-867-5
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.