Orders LinkContactUsLinkComponent
AMMONIUM HYDROXIDE, 0.1N STAND SOLUTION 1 * 4 L
AMMONIUM HYDROXIDE, 0.1N STAND SOLUTION 1 * 4 L
Katalog # 35576.K7
Leverandør:   Thermo Fisher Scientific
AMMONIUM HYDROXIDE, 0.1N STAND SOLUTION 1 * 4 L
Katalog # 35576.K7
CAS nummer:  
AMMONIUM HYDROXIDE, 0.1N STAND SOLUTION 1 * 4 L
CAS nummer:  
Price
1.770,00 kr
Log ind for tilgængelighed
1 * 4 L