Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyre standardised solution 0.1 N

Eddikesyre standardised solution 0.1 N

Glass bottle, 1 L
Katalog # 35567.K2
Leverandør:   Thermo Fisher Scientific
Eddikesyre standardised solution 0.1 N
Katalog # 35567.K2
CAS nummer:  64-19-7
Eddikesyre standardised solution 0.1 N
Eddikesyre standardised solution 0.1 N

Glass bottle, 1 L

CAS nummer:  64-19-7
Price
481,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Molecular Formula: CH3CO2H Formula Weight: 60.05 Physical Form: Liquid MDL No.: MFCD00036152

Synonymer:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS nummer:64-19-7
  • Formel:H₃CCOOH
  • MDL nummer:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-580-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.